missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress (S,R,S)-AHPC-PEG2-NH2 hydrochloride
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
10mg
1g
25mg
2g
500mg
50mg
Specifications
Specifications
| Molecular Weight (g/mol) | 612.18 g/mol |
| CAS | 2097973-72-1 |
| Content And Storage | 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture) |
| Shipping Condition | Room temperature |
| Quantity | 1 g |
| Synonym | VH032-PEG2-NH2 hydrochloride ∣ VHL Ligand-Linker Conjugates 3 hydrochloride ∣ E3 ligase Ligand-Linker Conjugates 6 |
| Solubility | H2O : 100 mg/mL (163.35 mM; Need ultrasonic) ∣ DMSO : 100 mg/mL (163.35 mM; Need ultrasonic) |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction