missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress (S,R,S)-AHPC-PEG2-NH2
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
Specifications
Specifications
| Molecular Weight (g/mol) | 575.72 g/mol |
| CAS | 2010159-60-9 |
| Content And Storage | Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Quantity | 100 mg |
| Synonym | VH032-PEG2-NH2 ∣ VHL Ligand-Linker Conjugates 3 ∣ E3 ligase Ligand-Linker Conjugates 6 Free Base |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction