Learn More
MedChemExpress (S,R,S)-AHPC-PEG1-N3
Specifications
Specifications
| Molecular Weight (g/mol) | 557.67 g/mol |
| CAS | 2101200-09-1 |
| Content And Storage | Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Quantity | 500 mg |
| Synonym | VH032-PEG1-N3 ∣ VHL Ligand-Linker Conjugates 9 ∣ E3 ligase Ligand-Linker Conjugates 3 |
| Solubility | DMSO : ≥ 100 mg/mL (179.32 mM) |