Learn More
(S)-(-)-Nicotine-d4, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-(-)-Nicotine pyridine-d4 |
| CAS | 284685-07-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 2H4 H10 N2 |
| Quantity | 10 mg |
| Synonym | 5-[(2S)-1-methyl-2-pyrrolidinyl]pyridine-2,3,4,6-d4, (S)-(-)-Nicotine-d4, (S)-(-)-Nicotine-D4 (pyridine-D4) |
| InChI Formula | InChI=1S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10H,3,5,7H2,1H3/t10-/m0/s1/i2D,4D,6D,8D |
| SMILES | [2H]c1nc([2H])c([C@@H]2CCCN2C)c([2H])c1[2H] |
| IUPAC Name | 2,3,4,6-tetradeuterio-5-[(2S)-1-methylpyrrolidin-2-yl]pyridine |
| Molecular Weight (g/mol) | 166.26 |
| Show More |