Learn More
(S)-N-(3-Chloro-4-fluorophenyl)-6-nitro-7-(tetrahydrofuran-3-yloxy)quinazolin-4-amine, TRC
Product Code. 30366404
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366404 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30366404 Supplier Toronto Research Chemicals Supplier No. TRCC367720500MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (S)-N-(3-Chloro-4-fluorophenyl)-6-nitro-7-(tetrahydrofuran-3-yloxy)quinazolin-4-amine
CAS 314771-88-5
Molecular Formula C18 H14 Cl F N4 O4
Quantity 500 mg
Synonym Afatinib Impurity 47
InChI Formula InChI=1S/C18H14ClFN4O4/c19-13-5-10(1-2-14(13)20)23-18-12-6-16(24(25)26)17(7-15(12)21-9-22-18)28-11-3-4-27-8-11/h1-2,5-7,9,11H,3-4,8H2,(H,21,22,23)
SMILES [O-][N+](=O)c1cc2c(Nc3ccc(F)c(Cl)c3)ncnc2cc1OC4CCOC4
IUPAC Name N-(3-chloro-4-fluorophenyl)-6-nitro-7-(oxolan-3-yloxy)quinazolin-4-amine
Molecular Weight (g/mol) 404.78
Formula Weight 404.069