Learn More
(S)-N-(3-Chloro-4-fluorophenyl)-6-nitro-7-(tetrahydrofuran-3-yloxy)quinazolin-4-amine, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-N-(3-Chloro-4-fluorophenyl)-6-nitro-7-(tetrahydrofuran-3-yloxy)quinazolin-4-amine |
| CAS | 314771-88-5 |
| Molecular Formula | C18 H14 Cl F N4 O4 |
| Quantity | 1 g |
| Synonym | Afatinib Impurity 47 |
| InChI Formula | InChI=1S/C18H14ClFN4O4/c19-13-5-10(1-2-14(13)20)23-18-12-6-16(24(25)26)17(7-15(12)21-9-22-18)28-11-3-4-27-8-11/h1-2,5-7,9,11H,3-4,8H2,(H,21,22,23) |
| SMILES | [O-][N+](=O)c1cc2c(Nc3ccc(F)c(Cl)c3)ncnc2cc1OC4CCOC4 |
| IUPAC Name | N-(3-chloro-4-fluorophenyl)-6-nitro-7-(oxolan-3-yloxy)quinazolin-4-amine |
| Molecular Weight (g/mol) | 404.78 |
| Formula Weight | 404.069 |