Learn More
(S)-Linezolid-d3 (acetamide-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Linezolid-d3 |
| CAS | 1127120-38-0 |
| Molecular Formula | C16 D3 H17 F N3 O4 |
| Quantity | 0.01 g |
| Synonym | 2,2,2-Trideuterio-N-[[(5 S)-3-(3-fluoro-4-morpholin-4-ylphenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl]acetamide, N-[[(5 S)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]acetamide-2,2,2-d3, Linezolid-D3 (acetyl-D3), (S)-Linezolid-d3, (S)-Linezolid-d3 (acetamide-d3) |
| InChI Formula | InChI=1 S/C16H20FN3O4/c1-11(21)18-9-13-10-20(16(22)24-13)12-2-3-15(14(17)8-12)19-4-6-23-7-5-19/h2-3,8,13 H,4-7,9-10H2,1H3,(H,18,21)/t13-/m0/s1/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C(=O)NC[C@H]1 CN(C(=O)O1)c2ccc(N3CCOCC3)c(F)c2 |
| IUPAC Name | 2,2,2-trideuterio-N-[[(5 S)-3-(3-fluoro-4-morpholin-4-ylphenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl]acetamide |
| Molecular Weight (g/mol) | 340.365 |
| Formula Weight | 340.163 g/mol |
| Show More |