Learn More
(S)-4-Nitrophenyl 1-Phenyl-3,4-dihydroisoquinoline-2(1H)-carboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-4-Nitrophenyl 1-Phenyl-3,4-dihydroisoquinoline-2(1H)-carboxylate |
| CAS | 1229227-22-8 |
| Molecular Formula | C22H18N2O4 |
| Quantity | 1 g |
| Synonym | (1S)-3,4-Dihydro-1-phenyl-2(1H)-isoquinolinecarboxylic Acid 4-Nitrophenyl Ester, p-Nitrophenyl (S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline-2-cate |
| SMILES | OC(=O)/C=C/c1c(C=O)cccc1 |
| Molecular Weight (g/mol) | 374.39 |