Learn More
(S)-3-Hydroxypyrrolidine, TRC
Product Code. 30390488
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30390488 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30390488 Supplier Toronto Research Chemicals Supplier No. TRCH953668250MG
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (3S)-3-Hydroxypyrrolidine
CAS 100243-39-8
Molecular Formula C4 H9 N O
Quantity 250 mg
Synonym 3-Pyrrolidinol, (3S)- (9CI, ACI), (3S)-3-Pyrrolidinol (ACI), 3-Pyrrolidinol, (S)- (ZCI), (+)-3-Pyrrolidinol, (3S)-3-Hydroxypyrrolidine, (3S)-Pyrrolidinol, (S)-(+)-3-Pyrrolidinol, (S)-3-Hydroxypyrrolidine, (S)-3-Pyrrolidinol, 3-(S)-Hydroxypyrrolidine
InChI Formula InChI=1S/C4H9NO/c6-4-1-2-5-3-4/h4-6H,1-3H2/t4-/m0/s1
SMILES O[C@H]1CCNC1
IUPAC Name (3S)-pyrrolidin-3-ol
Molecular Weight (g/mol) 87.12
Formula Weight 87.068