Learn More
(S)-3-(Boc-amino)-3-phenylpropionic acid, 95%
Product Code. p-7034389
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458597 250 mg 250mg
15438597 1 g 1g
15448597 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15458597 Supplier Thermo Scientific Alfa Aesar Supplier No. H52072.MD
Add to basket
CAS: 103365-47-5 | C14H19NO4 | 265.309 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 103365-47-5
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.309
MDL Number MFCD01860892
InChI Key JTNQFJPZRTURSI-NSHDSACASA-N
Synonym boc-s-3-amino-3-phenylpropionic acid, s-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, s-3-n-boc-amino-beta-phenylalanine, s-n-boc-3-amino-3-phenylpropanoic acid, boc-d-beta-phe-oh, boc-d-beta-phenylalanine, s-boc-beta3-phenylalanine, boc-s-phenylalanine, 3s-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, s-3-boc-amino-3-phenylpropionic acid
PubChem CID 2734490
IUPAC Name (3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC=CC=C1
Specifications
Quantity 250 mg
Beilstein 4695945
Formula Weight 265.31
Percent Purity 95%
Chemical Name or Material (S)-3-(Boc-amino)-3-phenylpropionic acid