Learn More
(S)-3-Acetyl-3,4-dihydro-3,5,12-trihydroxy-10-methoxy-1,6,11(2H)-naphthacenetrione (90%), TRC
Specifications
Specifications
| Chemical Name or Material | (S)-3-Acetyl-3,4-dihydro-3,5,12-trihydroxy-10-methoxy-1,6,11(2H)-naphthacenetrione |
| CAS | 32469-41-3 |
| Molecular Formula | C21H16O8 |
| Quantity | 1 mg |
| Synonym | (S)-3-Acetyl-3,5,12-trihydroxy-10-methoxy-3,4-dihydrotetracene-1,6,11(2H)-trione |
| InChI Formula | InChI=1S/C21H16O8/c1-8(22)21(28)6-10-13(11(23)7-21)19(26)16-15(18(10)25)17(24)9-4-3-5-12(29-2)14(9)20(16)27/h3-5,25-26,28H,6-7H2,1-2H3/t21-/m0/s1 |
| SMILES | COc1cccc2C(=O)c3c(O)c4C[C@](O)(CC(=O)c4c(O)c3C(=O)c12)C(=O)C |
| IUPAC Name | (3S)-3-acetyl-3,5,12-trihydroxy-10-methoxy-2,4-dihydrotetracene-1,6,11-trione |
| Molecular Weight (g/mol) | 396.35 |
| Formula Weight | 396.0845 |