missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Methyl-2-propanesulfinamide, 97%

Product Code. p-7029629
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11426210

Brand: Thermo Scientific Alfa Aesar H27115.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 343338-28-3 | C4H11NOS | 121.198 g/mol

(S)-(-)-2-Methyl-2-propanesulfinamide is used in Suzuki reaction. It is also employed as a reagent for synthesizing chiral amines. It acts as a chiral auxiliary used in an asymmetric synthesis of trifluoroethylamines by conversion of trifluoroacetaldehyde to a chiral imine. It is also involved in the transformation of P,N-sulfinyl imine ligands through condensation with aldehydes and ketones, which can undergo iridium-catalyzed asymmetric hydrogenation of olefins. Further, it serves as a reagent for the preparation of chemicals and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-2-Methyl-2-propanesulfinamide is used in Suzuki reaction. It is also employed as a reagent for synthesizing chiral amines. It acts as a chiral auxiliary used in an asymmetric synthesis of trifluoroethylamines by conversion of trifluoroacetaldehyde to a chiral imine. It is also involved in the transformation of P,N-sulfinyl imine ligands through condensation with aldehydes and ketones, which can undergo iridium-catalyzed asymmetric hydrogenation of olefins. Further, it serves as a reagent for the preparation of chemicals and pharmaceutical intermediates.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 343338-28-3
Molecular Formula C4H11NOS
Molecular Weight (g/mol) 121.198
MDL Number MFCD05861480
InChI Key CESUXLKAADQNTB-ZETCQYMHSA-N
Synonym s---2-methyl-2-propanesulfinamide, s-2-methylpropane-2-sulfinamide, s-tert-butanesulfinamide, s---tert-butanesulfinamide, s---t-butylsulfinamide, s---tert-butylsulfinamide, s-2-methyl-2-propanesulfinamide, s-tert-butylsulfinamide, s---t-butylmethylsulfinamide, s-t-butyl sulfinamide
PubChem CID 11355477
IUPAC Name 2-methylpropane-2-sulfinamide
SMILES CC(C)(C)S(=O)N

Specifications

Melting Point 97°C to 101°C
Quantity 1 g
Formula Weight 121.2
Percent Purity 97%
Chemical Name or Material (S)-(-)-2-Methyl-2-propanesulfinamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.