Learn More
(S)-2-(Dimethoxymethyl)-1-pyrrolidinecarboxylic Acid 1,1-Dimethylethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-2-(Dimethoxymethyl)-1-pyrrolidinecarboxylic Acid 1,1-Dimethylethyl Ester |
| CAS | 110562-19-1 |
| Molecular Formula | C12H23NO4 |
| Quantity | 5 mg |
| Synonym | (S)-t-Butyl 2-(dimethoxymethyl)pyrrolidine-1-carboxylate |
| InChI Formula | InChI=1S/C12H23NO4/c1-12(2,3)17-11(14)13-8-6-7-9(13)10(15-4)16-5/h9-10H,6-8H2,1-5H3/t9-/m0/s1 |
| SMILES | COC(OC)[C@@H]1CCCN1C(=O)OC(C)(C)C |
| IUPAC Name | tert-butyl (2S)-2-(dimethoxymethyl)pyrrolidine-1-carboxylate |
| Molecular Weight (g/mol) | 245.32 |
| Formula Weight | 245.1627 |