Learn More
(S)-2-Bromobutyric Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-2-Bromobutyric Acid |
| CAS | 32659-49-7 |
| Molecular Formula | C4H7BrO2 |
| Quantity | 2 g |
| Synonym | (S)-2-Bromobutanoic Acid, L-2-Bromobutanoic Acid, L-2-Bromobutyric Acid, L-alpha-Bromobutanoic Acid |
| InChI Formula | InChI=1S/C4H7BrO2/c1-2-3(5)4(6)7/h3H,2H2,1H3,(H,6,7)/t3-/m0/s1 |
| SMILES | CC[C@H](Br)C(=O)O |
| IUPAC Name | (2S)-2-bromobutanoic acid |
| Molecular Weight (g/mol) | 167 |
| Formula Weight | 165.9629 |