Learn More
(S)-2-Bromobutyric Acid, TRC
Product Code. 30329326
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30329326 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30329326 Supplier Toronto Research Chemicals Supplier No. TRCB545005500MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (S)-2-Bromobutyric Acid
CAS 32659-49-7
Molecular Formula C4H7BrO2
Quantity 500 mg
Synonym (S)-2-Bromobutanoic Acid, L-2-Bromobutanoic Acid, L-2-Bromobutyric Acid, L-alpha-Bromobutanoic Acid
InChI Formula InChI=1S/C4H7BrO2/c1-2-3(5)4(6)7/h3H,2H2,1H3,(H,6,7)/t3-/m0/s1
SMILES CC[C@H](Br)C(=O)O
IUPAC Name (2S)-2-bromobutanoic acid
Molecular Weight (g/mol) 167
Formula Weight 165.9629