Learn More
(S)-1-(Thiophen-2-yl)ethanol, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-1-(Thiophen-2-yl)ethanol |
| CAS | 27948-39-6 |
| Molecular Formula | C6 H8 O S |
| Quantity | 100 mg |
| Synonym | (alphaS)-alpha-Methyl-2-thiophenemethanol, (S)-1-(2-Thienyl)ethanol |
| InChI Formula | InChI=1S/C6H8OS/c1-5(7)6-3-2-4-8-6/h2-5,7H,1H3/t5-/m0/s1 |
| SMILES | C[C@H](O)c1cccs1 |
| IUPAC Name | (1S)-1-thiophen-2-ylethanol |
| Molecular Weight (g/mol) | 128.192 |
| Formula Weight | 128.03 |