Learn More
(S)-1-[(RP)-2-(Diphenylphosphino)ferrocenyl]ethyldi-tert-butylphosphine, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-1-[(RP)-2-(Diphenylphosphino)ferrocenyl]ethyldi-tert-butylphosphine |
| CAS | 277306-29-3 |
| Molecular Formula | C32H40FeP2 |
| Quantity | 25 mg |
| Synonym | (2S)-1-[(1S)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(diphenylphosphino)ferrocene |
| InChI Formula | InChI=1S/C27H35P2.C5H5.Fe/c1-21(29(26(2,3)4)27(5,6)7)24-19-14-20-25(24)28(22-15-10-8-11-16-22)23-17-12-9-13-18-23;1-2-4-5-3-1;/h8-21H,1-7H3;1-5H;/q2*-1;+2/t21-;;/m0../s1 |
| SMILES | CC(C)(C)P(C(C)(C)C)[C@@H](C)C12=[CH](C3([H])=C45[H])[Fe+2]1346789([CH-]%10[CH]6=[CH]7[CH]8=[CH]9%10)[C-]25P(C%11=CC=CC=C%11)C%12=CC=CC=C%12 |
| Molecular Weight (g/mol) | 542.45 |
| Formula Weight | 542.2 |