Learn More
(S)-1-Pyridin-2-yl-ethylamine, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-1-Pyridin-2-yl-ethylamine |
| CAS | 27854-90-6 |
| Molecular Formula | C7 H10 N2 |
| Quantity | 1 g |
| Synonym | 2-Pyridinemethanamine','α-methyl-','(αS)- (9CI','ACI), (αS)-α-Methyl-2-pyridinemethanamine (ACI), 2-Pyridinemethanamine','α-methyl-','(S)- (ZCI), Pyridine','2-(1-aminoethyl)-','(S)-(-)- (8CI), (-)-(S)-1-(2-Pyridyl)ethanamine, (1S)-1-(2-Pyridyl)ethanamine, (1S)-1-(Pyridin-2-yl)ethan-1-amine, (1S)-1-Pyridin-2-ylethanamine, (S)-1-(2-Pyridyl)ethylamine, (S)-1-(Pyridin-2-yl)ethan-1-amine, (S)-1-(Pyridin-2-yl)ethanamine, (S)-1-(Pyridin-2-yl)ethylamine, (S)-2-(1-Aminoethyl)pyridine, 2-[(S)-1-Aminoethyl]pyridine |
| InChI Formula | InChI=1S/C7H10N2/c1-6(8)7-4-2-3-5-9-7/h2-6H,8H2,1H3/t6-/m0/s1 |
| SMILES | C[C@H](N)c1ccccn1 |
| IUPAC Name | (1S)-1-(2-pyridyl)ethanamine |
| Molecular Weight (g/mol) | 122.168 |
| Formula Weight | 122.0844 |