Learn More
(S)-1-Chloro-3-((3-fluoro-4-morpholinophenyl)amino)propan-2-ol, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-1-Chloro-3-((3-fluoro-4-morpholinophenyl)amino)propan-2-ol |
| CAS | 1610690-08-8 |
| Molecular Formula | C13H18ClFN2O2 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C13H18ClFN2O2/c14-8-11(18)9-16-10-1-2-13(12(15)7-10)17-3-5-19-6-4-17/h1-2,7,11,16,18H,3-6,8-9H2/t11-/m1/s1 |
| SMILES | O[C@H](CCl)CNc1ccc(N2CCOCC2)c(F)c1 |
| IUPAC Name | (2S)-1-chloro-3-(3-fluoro-4-morpholin-4-ylanilino)propan-2-ol |
| Molecular Weight (g/mol) | 288.75 |
| Formula Weight | 288.1041 |