missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-Boc-2-isobutylpiperazine, 97%

Product Code. p-7034712
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499127 250 mg 250mg
15489127 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499127 Supplier Thermo Scientific Alfa Aesar Supplier No. H52409.MD
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 674792-06-4 | C13H26N2O2 | 242.363 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 674792-06-4
Molecular Formula C13H26N2O2
Molecular Weight (g/mol) 242.363
MDL Number MFCD03787916
InChI Key WXGGVOBNOVOVAM-NSHDSACASA-N
Synonym s-1-boc-2-isobutyl-piperazine, s-tert-butyl 2-isobutylpiperazine-1-carboxylate, s-1-boc-2-isobutylpiperazine, tert-butyl 2s-2-2-methylpropyl piperazine-1-carboxylate, s-1-n-boc-isobutylpiperazine, s-n-boc-2-isobutylpiperazine, tert-butyl 2s-2-isobutylpiperazine-1-carboxylate, s-1-n-boc-2-isobutylpiperazine, 2s-1-n-boc-2-isobutylpiperazine, 2s-1-tert-butoxycarbonyl-2-isobutylpiperazine
PubChem CID 17750437
IUPAC Name tert-butyl (2S)-2-(2-methylpropyl)piperazine-1-carboxylate
SMILES CC(C)CC1CNCCN1C(=O)OC(C)(C)C

Specifications

Quantity 250 mg
Formula Weight 242.36
Percent Purity 97%
Chemical Name or Material (S)-1-Boc-2-isobutylpiperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.