missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-(4-Chlorophenyl)ethylamine, prd by BASF AG, 95+% (sum of isomers), 95% e., ACROS Organics™

Product Code. 12736893 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12736893 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12736893 Supplier Thermo Scientific Acros Supplier No. 370660050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4187-56-8
Color Colorless to Yellow
Density 1.1150g/mL
Quantity 5 g
Infrared Spectrum Authentic
Physical Form Liquid
Refractive Index 1.5410 to 1.5430 (20°C, 589nm)
Molecular Formula C8H10ClN
Linear Formula ClC6H4CH(CH3)NH2
MDL Number MFCD00671640
Synonym s-1-4-chlorophenyl ethylamine, s-1-4-chlorophenyl ethanamine, 1s-1-4-chlorophenyl ethanamine, s-4-chloro-alpha-methylbenzylamine, s---1-4-chlorophenyl ethylamine, 1s-1-4-chlorophenyl ethan-1-amine, pubchem15221, s-4-chloro-, a-methylbenzylamine, 1s-1-4-chlorophenyl ethylamine, s-1-4-chloro-phenyl ethylamine
InChI Key PINPOEWMCLFRRB-LURJTMIESA-N
SMILES CC(C1=CC=C(C=C1)Cl)N
IUPAC Name (1S)-1-(4-chlorophenyl)ethanamine
Molecular Weight (g/mol) 155.625
PubChem CID 1550852
Formula Weight 155.63
Percent Purity ≥95.0% (sum of isomers)
Chemical Name or Material (S)-1-(4-Chlorophenyl)ethylamine, produced by BASF AG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.