missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-(3-Chlorophenyl)ethylamine, ChiPros™, 98%, ee 98+%, Thermo Scientific™

Product Code. p-7030100
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11496590

Brand: Thermo Scientific Alfa Aesar H27637.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
As active pharmaceutical drugs and intermediates possess different biological activities.

Notes
Moisture Sensitive. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air. Refrigerated (2-4°C).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68297-62-1
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.63
MDL Number MFCD06761823
InChI Key DQEYVZASLGNODG-UHFFFAOYNA-N
Synonym s-1-3-chlorophenyl ethanamine, s-1-3-chlorophenyl ethylamine, 1s-1-3-chlorophenyl ethanamine, 1s-1-3-chlorophenyl ethan-1-amine, s-3-chloro-, a-methylbenzylamine, s---1-3-chlorophenyl ethylamine, s-1-3-chlorophenyl ethylamine, chipros, benzenemethanamine, 3-chloro-, a-methyl-,, as, s-3-chloro-alpha-methylbenzylamine, chipros r , produced by basf
PubChem CID 2507691
IUPAC Name (1S)-1-(3-chlorophenyl)ethanamine
SMILES CC(N)C1=CC(Cl)=CC=C1

Specifications

Boiling Point 112°C (13 mmHg)
Quantity 1 g
UN Number UN2735
Formula Weight 155.63
Percent Purity 98%
Assay ee ≥98%
Chemical Name or Material (S)-1-(3-Chlorophenyl)ethylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.