Learn More
VACUUBRAND™ RZ 2.5 Rotary Vane Pump
Product Code. p-8008759
Click to view available options
Model:
RZ 2.5 Package
Plug Type:
CEE
Unit Size:
Each
Product Code. 10594322
Brand: VACUUBRAND™ 20698029
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use for a wide range of laboratory and process applications that require low ultimate vacuum at medium gas flow rate Vacuubrand™ RZ 2.5 Rotary Vane Pump are high-performance rotary vane pumps with extra compact design and low weight.
High flow rates even at vacuum levels approaching ultimate vacuum
High water vapor tolerance due to efficient gas ballast; very good ultimate vacuum even with gas ballast
Vacuum-tight at switch-off; external anti-suckback valve not needed large oil volume
Long intervals between oil changes ease of maintenance due to telescopic design
TRUSTED_SUSTAINABILITY
Specifications
Max. Vacuum 1.5 x 10-3 torr (without Gas Ballast), 7.5 x 10-3 torr (with Gas Ballast)
Horsepower 0.18 hp
Includes CEE Plug, Isolation Valve VS 16, Manual, Oil Mist Filter
Motor Speed 150 to 1800 rpm
Certifications/Compliance CAN/CSA C22.2 61010-1-12, CE, DIN EN 1012-2:2011, DIN EN 12100:2001, DIN EN 50581:2013, DIN EN 61010-1:2011, DIN EN 61326-1:2013, IEC 61010-1:2010, TUV, UL 61010A 61010-1:2012
Electrical Requirements 230 V 50/60 Hz
Dimensions (L x W x H) 316 x 125 x 190 mm
Model RZ 2.5 Package
Noise Level 43 dBA
Plug Type CEE
Temperature (Metric) Storage -10°C to +60°C
Temperature (English) Max. 104°F
Temperature (Metric) Max 40°C
Temperature (Metric) Min. 12°C
Flow Rate 1.4/1.7 CFM
Frequency 50/60 Hz
Overall Height (Metric) 190.5 mm
Overall Length (Metric) 314.96 mm
Overall Width (Metric) 124.46 mm
Pump Type Rotary Vane Vacuum Pump
Enclosure Rating IP40
Resistance Properties Chemical-Resistant
Temperature (English) Min 54°F
Oil Capacity 0.11 to 0.3 qt.
Applications Laboratory
Oil Type Oil-Sealed
No. of Stages 2
Maximum Pressure 16 psi
Voltage 230 V