missing translation for 'onlineSavingsMsg'
Learn More

Ryanodine Receptor Antibody (34C) - BSA Free, Novus Biologicals™

Product Code. 18271051 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18271051 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18271051 Supplier Novus Biologicals Supplier No. NB300543

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 11 publications

Ryanodine Receptor Monoclonal specifically detects Ryanodine Receptor in Human, Mouse, Rat, Amphibian, Bovine, Canine, Fish, Ferret, Mink, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Block/Neutralize.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ryanodine Receptor
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Neutralization
Classification Monoclonal
Clone 34C
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:1000, Neutralization
Gene Accession No. Q90984
Gene Alias ARVC2, ARVD2, cardiac muscle ryanodine receptor-calcium release channel, cardiac-type ryanodine receptor, CCO, central core disease of muscle, islet-type ryanodine receptor, kidney-type ryanodine receptor, MHS, MHS1, PPP1R137, protein phosphatase 1, regulatory subunit 137, ryanodine receptor 1, ryanodine receptor 1 (skeletal), ryanodine receptor 2, ryanodine receptor 2 (cardiac), RYDR, RYR, RYR-1, RYR-2, sarcoplasmic reticulum calcium release channel, Skeletal muscle calcium release channel, skeletal muscle ryanodine receptor, SKRR, type 1-like ryanodine receptor, type 2 ryanodine receptor, VTSIP
Gene Symbols RYR1
Host Species Mouse
Immunogen Partially purified chicken pectoral muscle ryanodine receptor.
Molecular Weight of Antigen 565 kDa
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6261
Test Specificity This antibody detects ryanodine receptor Ryanodine Receptor-1 and Ryanodine Receptor-2 isoforms in amphibian, chicken, bovine, canine, fish, human, non-human primate, mouse, rabbit, mink, rat and sheep tissues. In chicken, it detects the alpha, beta and cardiac isoforms. This antibody also detects Ryanodine Receptor-3 in mouse cells. In frog, it detects the alpha and beta isoforms and in fish it detects the alpha isoform.
Target Species Human, Mouse, Rat, Amphibian, Bovine, Canine, Fish, Ferret, Mink, Primate, Rabbit, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.