missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RWJ 56110, Tocris Bioscience™
Description
Selective protease-activated receptor-1 (PAR1) antagonist; displays no activity at PAR2, PAR3, or PAR4 subtypes. Blocks thrombin-induced platelet aggregation and activation of MAPK in HUVECs. Also inhibits angiogenesis in a chick embryo angiogenesis model in vivo.
Specifications
Specifications
| CAS | 2387505-58-8 |
| Molecular Formula | C41H45Cl4F2N7O3 |
| Quantity | 1 mg |
| Synonym | ?s-n-1s-3-amino-1-phenylmethyl amino carbonyl propyl-?-1-2,6-dichlorophenyl methyl-3-1-pyrrolidinylmethyl-1h-indol-6-yl amino carbonyl amino-3,4-difluoro-benzenepropanamide |
| InChI Key | MGZSVSHDIOPSBO-ZEUUMAKDSA-N |
| SMILES | Cl.Cl.NCC[C@H](NC(=O)[C@H](CC1=CC(F)=C(F)C=C1)NC(=O)NC1=CC=C2C(CN3CCCC3)=CN(CC3=C(Cl)C=CC=C3Cl)C2=C1)C(=O)NCC1=CC=CC=C1 |
| IUPAC Name | (2S)-4-amino-N-benzyl-2-[(2S)-2-[({1-[(2,6-dichlorophenyl)methyl]-3-[(pyrrolidin-1-yl)methyl]-1H-indol-6-yl}carbamoyl)amino]-3-(3,4-difluorophenyl)propanamido]butanamide dihydrochloride |
| Molecular Weight (g/mol) | 863.65 |
| PubChem CID | 90488773 |
| Formula Weight | 863.65 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?