missing translation for 'onlineSavingsMsg'
Learn More

Bolle Safety RUSH+ 2.0 XP Safety Eyewear with Platinum Anti-fog, Anti-scratch Coating, Foam and Strap Kit

Product Code. 31810809 Shop All Bollé Safety Products
Change view
Click to view available options
Lens Tint:
Clear
Copper
Smoke
Size:
Medium/Large
Small
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Tint Size Unit Size
31810809 Copper Medium/Large Each
31810801 Clear Medium/Large Each
31810784 Smoke Medium/Large Each
31810774 Clear Small Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31810809 Supplier Bolle Safety Supplier No. RUSXMN30E
  • ONLINE EXCLUSIVE PRICE
    19.15 €
    17.23 € / Pack of 1
    Save 1.92 €
    10% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Delivers advanced eye protection for harsh environments with added sealing and support, ensuring a secure fit and clear vision.

  • Advanced protection for harsh environments: Built on the trusted RUSH+ 2.0 platform, the XP version enhances protection in demanding and high-risk conditions.
  • Foam gasket for extended sealing: Replaceable foam gasket improves protection against dust, particles, and debris, making it ideal for more exposed environments.
  • Adjustable strap for secure fit: Ensures optimal stability during movement and intensive tasks, improving safety and wearer confidence.
  • Wide field of vision: Large lens design enhances peripheral visibility, increasing awareness and reducing workplace hazards.
  • PLATINUM coating technology: Premium anti-fog and anti-scratch coating guarantees long-lasting clarity and performance in challenging conditions.
  • Enhanced comfort and support: Combines ergonomic design with additional support features to maintain comfort during extended wear.
  • Improved protection against ingress: Foam seal and secure fit help reduce gaps, limiting entry of dust and airborne particles.
  • Durable and long-lasting: Designed for intensive use, reducing replacement frequency and total cost of ownership.
  • Eco-conscious design: Made with 34% recycled materials, supporting sustainability initiatives and reducing environmental impact.
  • Boosts worker compliance: Comfortable, stable, and protective design encourages consistent use in demanding environments.
  • Versatile industrial application: Ideal for construction, heavy industry, maintenance, and environments requiring enhanced eye protection.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN ISO 16321-1
Lens Type C-Rated (Impact), GL1 Rated
Lens Coating Platinum, Anti-fog, Anti-scratch
Lens Tint Copper
Product Type Safety Glasses
Frame Color Gray, Black
Size Medium/Large
Material Polycarbonate, TPR
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate, TPR
UV Protection UV380
Frame Type Frameless
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.