missing translation for 'onlineSavingsMsg'
Learn More

Rucaparib, MedChemExpress

Product Code. p-200101637
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494068 10 mg 10mg
16455046 100 mg 100mg
16450427 50 mg 50mg
16416777 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494068 Supplier Medchem Express Supplier No. HY10617A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rucaparib (AG014699) is an orally active, potent inhibitor of PARP proteins (PARP-1, PARP-2 and PARP-3) with a Ki of 1.4 nM for PARP1. Rucaparib is a modest hexose-6-phosphate dehydrogenase (H6PD) inhibitor. Rucaparib has the potential for castration-resistant prostate cancer (CRPC) research.

Rucaparib (AG014699) is an orally active, potent inhibitor of PARP proteins (PARP-1, PARP-2 and PARP-3) with a Ki of 1.4 nM for PARP1. Rucaparib is a modest hexose-6-phosphate dehydrogenase (H6PD) inhibitor. Rucaparib has the potential for castration-resistant prostate cancer (CRPC) research.

Specifications

Chemical Name or Material Rucaparib
CAS 283173-50-2
Color Green Yellow
Purity Grade Notes Research
Molecular Formula C19H18FN3O
Quantity 10 mg
Synonym AG014699 PF-01367338
Solubility Information DMSO : 25 mg/mL (77.31 mM; ultrasonic and adjust pH to 4 with HCl) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES FC1=CC2=C3C(CCNC2=O)=C(C4=CC=C(CNC)C=C4)NC3=C1
Molecular Weight (g/mol) 323.36
Formula Weight 323.36
Percent Purity 99.27%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.