missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Rucaparib camsylate
GREENER_CHOICE

Product Code. 18779816
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18779816 50 mg 50mg
18708776 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18779816 Supplier Tocris Bioscience™ Supplier No. 6230/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PARP inhibitor

Rucaparib camsylate is a PARP inhibitor. Chemosensitizes colorectal cancer cells to Temozolomide (Cat. No. 2706) and Topotecan (Cat. No. 4562) in vitro and in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-Fluoro-1,3,4,5-tetrahydro-2-[4-[(methylamino)methyl]phenyl]-6 H-pyrrolo[4,3,2,ef][2 ]benzazepin-6-one (1 S,4 R)-7,7-dimethyl-2-oxobicyclo[2.2.1 ]heptane-1-methanesulfonic acid
CAS 1859053-21-6
Assay Percent Range >98%
Quantity 50 mg
Target Poly(ADP-ribose) Polymerase Inhibitors
Molecular Formula C19H18FN3O.C10H16O4S
Formula Weight Observed MW: 555.66
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.