missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Rucaparib camsylate

Description
Rucaparib camsylate is a PARP inhibitor. Chemosensitizes colorectal cancer cells to Temozolomide (Cat. No. 2706) and Topotecan (Cat. No. 4562) in vitro and in vivo.
Specifications
Specifications
| Chemical Name or Material | 8-Fluoro-1,3,4,5-tetrahydro-2-[4-[(methylamino)methyl]phenyl]-6 H-pyrrolo[4,3,2,ef][2 ]benzazepin-6-one (1 S,4 R)-7,7-dimethyl-2-oxobicyclo[2.2.1 ]heptane-1-methanesulfonic acid |
| CAS | 1859053-21-6 |
| Assay Percent Range | >98% |
| Quantity | 10 mg |
| Target | Poly(ADP-ribose) Polymerase Inhibitors |
| Molecular Formula | C19H18FN3O.C10H16O4S |
| Formula Weight | Observed MW: 555.66 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?