missing translation for 'onlineSavingsMsg'
Learn More

RuBi-Nicotine, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18722514
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18722514 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18722514 Supplier Tocris Bioscience Supplier No. 3855/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Caged nicotine; rapidly excitable by visible light

Ruthenium-bisbipyridine-caged nicotine, excitable by irradiation with visible light. Undergoes rapid photolysis to release nicotine (< 20 ns); exhibits a high quantum yield. Induces action potential propagation in Retzius neurons of leech ganglia with no detectable toxicity at a concentration of 1 mM.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1256362-30-7
Molecular Formula C40H44Cl2N8Ru
Quantity 10 mg
Synonym rubi-nicotine, bis 2,2'-bipyridine-n,n' bis s-nicotine-n1 ruthenium 2+ dichloride
InChI Key BQXVYFYPLMVART-ZPFSJBFKSA-L
SMILES CN1CCCC1C2=CN=CC=C2.CN1CCCC1C2=CN=CC=C2.C1=CC=NC(=C1)C2=CC=CC=N2.C1=CC=NC(=C1)C2=CC=CC=N2.[Cl-].[Cl-].[Ru+2]
IUPAC Name 3-[(2S)-1-methylpyrrolidin-2-yl]pyridine;2-pyridin-2-ylpyridine;ruthenium(2+);dichloride
Molecular Weight (g/mol) 808.818
PubChem CID 129316181
Formula Weight 808.81
Percent Purity >98%
Chemical Name or Material RuBi-Nicotine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.