Learn More
Cole-Parmer™ Rubbermaid™ NSF-Approved High-Heat Spatulas
Product Code. 11720459 Shop All Cole-Parmer Products
Click to view available options
Length (Metric):
41.91 cm
Unit Size:
Pack of 6
Product Code. 11720459
Brand: Cole-Parmer™ 0627835
Please to purchase this item. Need a web account? Register with us today!
Scooping, spooning and spreading have never been easier with these spoon-shaped spatulas.
  • The sturdy polypropylene (PP) handle is permanently bonded to a flexible rubber blade
  • Spatulas withstand temperatures to 212°F (100°C)
  • NSF-approved
Specifications
Certifications/Compliance NSF-Approved
Product Type High-Heat Spatula
Length (English) 16.5 in.
Material Polypropylene, Rubber
Length (Metric) 41.91 cm