missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Rubber Tubing, Red

Product Code. 10525002
Change view
Click to view available options
Diameter (Metric) Inner:
4 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
10525002 4 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10525002 Supplier Karl Hecht™ Supplier No. 41580004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Economical tubing with good elasticity retention. Karl Hecht™ Rubber Tubing, Red is useful for a variety of laboratory applications.

  • Red rubber construction.
  • Maintains elasticity.

Specifications

Diameter (Metric) Inner 4 mm
Material Rubber
Thickness (Metric) 1 mm
Color Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.