Learn More
RU 58841, MedChemExpress
Description
RU 58841 (PSK-3841) is a specific androgen receptor antagonist or anti-androgen. RU 58841 (PSK-3841) has a dramatic effect on hair regrowth.
Specifications
Specifications
| Chemical Name or Material | RU 58841 |
| Physical Form | Solid |
| CAS | 154992-24-2 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C17H18F3N3O3 |
| Synonym | PSK-3841 HMR-3841 |
| Solubility Information | DMSO : ≥ 100 mg/mL (270.75 mM) |
| SMILES | FC(F)(C1=C(C=CC(N2C(C(C)(N(C2=O)CCCCO)C)=O)=C1)C#N)F |
| Show More |