missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RTP1 Polyclonal Antibody

Description
This antibody is predicted to react with non-human primate based on sequence homology.
Specifically promotes functional cell surface expression of olfactory receptors, but not of other GPCRs.
Specifications
Specifications
| Antigen | RTP1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Rtp1 |
| Gene Accession No. | P59025, Q8C8C1 |
| Gene Alias | 3CxxC-type zinc finger protein 1; Gm1766; Gm604; receptor (chemosensory) transporter protein 1; receptor transporter protein 1; receptor transporting protein 1; receptor-transporting protein 1; RGD1560980; Rtp1; Z3CXXC1; zinc finger, 3CxxC-type 1 |
| Gene Symbols | Rtp1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?