Learn More
RTI-336, TRC
Specifications
Specifications
| Chemical Name or Material | 5-[(1S,3S,4S,5R)-3-(4-chlorophenyl)-8-methyl-8-azabicyclo[3.2.1]octan-4-yl]-3-(4-methylphenyl)-1,2-oxazole;hydrochloride |
| CAS | 204069-50-1 |
| Molecular Formula | C24 H25 Cl N2 O . H Cl |
| Quantity | 5 mg |
| Synonym | (1R,2S,3S,5S)-3-(4-Chlorophenyl)-8-methyl-2-[3-(4-methylphenyl)-5-isoxazolyl]-8-azabicyclo[3.2.1]octane Hydrochloride, [1R-(exo,exo)]-3-(4-Chlorophenyl)-8-methyl-2-[3-(4-methylphenyl)-5-isoxazolyl]-8-azabicyclo[3.2.1]octane Hydrochloride, RTI 336, RTI 4229-336, RTI-336 |
| InChI Formula | InChI=1S/C24H25ClN2O.ClH/c1-15-3-5-17(6-4-15)21-14-23(28-26-21)24-20(16-7-9-18(25)10-8-16)13-19-11-12-22(24)27(19)2;/h3-10,14,19-20,22,24H,11-13H2,1-2H3;1H/t19-,20+,22+,24-;/m0./s1 |
| SMILES | Cl.CN1[C@H]2CC[C@@H]1[C@H]([C@H](C2)c3ccc(Cl)cc3)c4onc(c4)c5ccc(C)cc5 |
| IUPAC Name | 5-[(1S,3S,4S,5R)-3-(4-chlorophenyl)-8-methyl-8-azabicyclo[3.2.1]octan-4-yl]-3-(4-methylphenyl)-1,2-oxazole;hydrochloride |
| Molecular Weight (g/mol) | 429.382 |
| Formula Weight | 428.142 |