Learn More
Tocris Bioscience™ RS 17053 hydrochloride
GREENER_CHOICE
Product Code. 18054482
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18054482
Brand: Tocris Bioscience™ 0985/10
Please to purchase this item. Need a web account? Register with us today!
α1A antagonist
α1A-adrenoceptor antagonist, with very high affinity for α1A receptors (pKi and pA2 estimates of 9.1 - 9.9) and a 30 - 100 fold selectivity over the α1B and α1D subtypes (pKi and pA2 estimates 7.7 - 7.8).
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 169505-93-5
Molecular Formula C24H30Cl2N2O2
Specifications
Quantity 10 mg
Formula Weight 449.42
Purity >98%
Molecular Weight (g/mol) 449.416
Chemical Name or Material RS 17053 hydrochloride