missing translation for 'onlineSavingsMsg'
Learn More

RS 17053 hydrochloride, MedChemExpress

Product Code. p-200099606
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16499866

Brand: Medchem Express HY101336

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RS 17053 hydrochloride is a potent and selective α1A adrenoceptor antagonist, with a pKi value of 9.1 in native cell membrane and a pA2 value of 9.8 in functional assays.

RS 17053 hydrochloride is a potent and selective α1A adrenoceptor antagonist, with a pKi value of 9.1 in native cell membrane and a pA2 value of 9.8 in functional assays.

Specifications

Chemical Name or Material RS 17053 hydrochloride
CAS 169505-93-5
Color White
Purity Grade Notes Research
Molecular Formula C24H30Cl2N2O2
Quantity 50 mg
Synonym RS-17053
Solubility Information DMSO : ≥ 125 mg/mL (278.14 mM)
SMILES [H]Cl.ClC1=CC2=C(NC=C2CC(C)(C)NCCOC3=CC=CC=C3OCC4CC4)C=C1
Molecular Weight (g/mol) 449.41
Formula Weight 449.41
Percent Purity 99.11%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.