missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RRP36 Polyclonal antibody specifically detects RRP36 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | RRP36 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | C6orf153, chromosome 6 open reading frame 153, dJ20C7.4, ribosomal RNA processing 36 homolog (S. cerevisiae), ribosomal RNA processing protein 36 homolog, RNA processing factor |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-259 of human RRP36 (NP_149103.1). MPGANYRAGAGAGAGARRPRGARDREEDGGGLEPAAVARDLLRGTSNMSFEELLELQSQVGTKTYKQLVAGNSPKKQASRPPIQNACVADKHRPLEMSAKIRVPFLRQVVPISKKVARDPRFDDLSGEYNPEVFDKTYQFLNDIRAKEKELVKKQLKKHLSGEEHEKLQQLLQRMEQQEMAQQERKQQQELHLALKQERRAQAQQGHRPYFLKKSEQRQLALAEKFKELKRSKKLENFLSRKRRRNAGKDRRHLPLSKE |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?