missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS4X Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
113.00 € - 363.00 €
Specifications
| Antigen | RPS4X |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18631575
|
Novus Biologicals
NBP3-21449-25ul |
25 μg |
113.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692735
|
Novus Biologicals
NBP3-21449-100ul |
100 μg |
363.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RPS4X Polyclonal antibody specifically detects RPS4X in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| RPS4X | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing | |
| PBS, pH 7.0 | |
| 6191 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CCG2Single copy abundant mRNA protein, DXS306, FLJ40595, ribosomal protein S4, X-linked, RPS4cell cycle gene 2, SCAR40S ribosomal protein S4, X isoform, SCR10ribosomal protein S4X isoform, single-copy abundant mRNA | |
| Produced in rabbits immunized with E. coli-derived Human RPS4X fragment | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title