missing translation for 'onlineSavingsMsg'
Learn More

RPS4X Antibody [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30174438
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30174438 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30174438 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP321449JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RPS4X Polyclonal antibody specifically detects RPS4X in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPS4X
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias CCG2Single copy abundant mRNA protein, DXS306, FLJ40595, ribosomal protein S4, X-linked, RPS4cell cycle gene 2, SCAR40S ribosomal protein S4, X isoform, SCR10ribosomal protein S4X isoform, single-copy abundant mRNA
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RPS4X fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 6191
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.