missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
15566-1-AP is a rabbit polyclonal antibody raised against full-length human RPS25.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S25E family of ribosomal proteins. It is located in the cytoplasm.
Specifications
Specifications
| Antigen | RPS25 |
| Applications | ELISA, Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 0.16 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RPS25 |
| Gene Accession No. | P62851 |
| Gene Symbols | RPS25 |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?