missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPP30 Polyclonal Antibody

Description
Recommended positive controls: 293T. Predicted reactivity: Mouse (89%), Rat (90%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RNase P, which consists of a complex of an RNA species (H1 RNA), POP1 (Processing of Precursor 1), POP5 (Processing of Precursor 5), and at least seven Rpps (including Rpp14, Rpp29, Rpp30 and Rpp38), removes the 5' leader sequences from precursor tRNA molecules. The structurally related RNase MRP plays an essential role in the formation of the 5' end of 5.8S rRNA. Both RNase P and RNase MRP are associated with Th/To ribonucleoproteins; Rpp30 and Rpp38 have specifically been implicated as Th autoantigens which contribute to the autoimmune disease systemic sclerosis.
Specifications
Specifications
| Antigen | RPP30 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.57 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | RPP30 |
| Gene Accession No. | P78346 |
| Gene Alias | Ribonuclease P protein subunit p30; ribonuclease P/MRP 30 subunit; ribonuclease P/MRP 30kDa subunit; ribonuclease P/MRP subunit; ribonuclease P/MRP subunit p30; ribonuclease P2; RNase P subunit 2; RNaseP protein p30; RNASEP2; RPP30; TSG15 |
| Gene Symbols | RPP30 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?