Learn More
Roxithromycin, MedChemExpress
Product Code. 16482169
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482169 1 g 1g
16460928 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16482169 Supplier Medchem Express Supplier No. HYB0435
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress Roxithromycin (RU-28965) is a semi-synthetic macrolide antibiotic.
Roxithromycin (RU-28965) is a semi-synthetic macrolide antibiotic.
Specifications
Chemical Name or Material Roxithromycin
Physical Form Solid
CAS 80214-83-1
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C41H76N2O15
For Use With (Application) COVID-19-immunoregulation
Synonym RU-28965
Solubility Information DMSO : ≥ 100 mg/mL (119.47 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@H]([C@H]([C@](O)(C[C@H](/C1=N\OCOCCOC)C)C)O[C@@](O[C@H](C)C[C@@H]2N(C)C)([H])[C@@H]2O)[C@]([C@H](C(O[C@@H]([C@@](O)([C@H](O)[C@H]1C)C)CC)=O)C)([H])O[C@@](O[C@@H](C)[C@@H]3O)([H])C[C@@]3(C)OC
Molecular Weight (g/mol) 837.05
Formula Weight 837.05
Percent Purity 98.0%
Grade Research