missing translation for 'onlineSavingsMsg'
Learn More

Roxithromycin, 97%

Product Code. 15497479
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497479 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15497479 Supplier Thermo Scientific Alfa Aesar Supplier No. J66213.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Semi-synthetic macrolide antibiotic. | CAS: 80214-83-1 | C41H76N2O15 | 837.06 g/mol

Used to block bacterial protein synthesis. Clinically, used to treat respiratory tract, urinary and soft tissue infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80214-83-1
Molecular Formula C41H76N2O15
Molecular Weight (g/mol) 837.06
MDL Number MFCD00214389
InChI Key RXZBMPWDPOLZGW-KMAKEOJNSA-N
Synonym roxithromycin
PubChem CID 133687104
IUPAC Name (3R,4S,5S,6R,7R,9R,10E,11S,12R,13S,14R)-6-{[4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-14-ethyl-7,12,13-trihydroxy-4-[(5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl)oxy]-3,5,7,9,11,13-hexamethyl-10-(2,4,7-trioxa-1-azaoctan-1-ylidene)-1-oxacyclotetradecan-2-one
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](OC2CC(C)(OC)C(O)C(C)O2)[C@H](C)[C@@H](OC2OC(C)CC(C2O)N(C)C)[C@](C)(O)C[C@@H](C)\C(=N/OCOCCOC)[C@H](C)[C@@H](O)[C@]1(C)O

Specifications

Melting Point 111°C to 118°C
Flash Point >110°C (230°F)
Packaging Glass bottle
Quantity 10 g
Solubility Information Soluble in water to 18μg/ml
Formula Weight 837.04
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Roxithromycin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.