Learn More
Gravis Round Wide Opening Glass, Screw-in Capsule
Product Code. p-7224414
Click to view available options
Color:
Amber
White
Unit Size:
Pack of 1560
Pack of 42
This item is not returnable. View return policy
Product Code. 17522015
Brand: Gravis B00454B
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Provides excellent storage for liquids and solids in the laboratory. Gravis Wide Neck Glass Bottles are to be sealed with waterproof screw caps.
  • Round Wide Opening Amber Glass
  • 250 mL Capacity
  • Made of Borosilicate Glass (3.3)
  • Spouted Cap Included
Specifications
Capacity (Metric) 250 mL
Color Amber
Material Borosilicate Glass (3.3)
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide
Shape Round
Closure Type Spouted Cap
Autoclavable Autoclavable