missing translation for 'onlineSavingsMsg'
Learn More

Rosmarinic Acid, TRC

Product Code. 17989454
Change view
Click to view available options
Quantity:
10 mg
50 mg
5 g
Unit Size:
10mg
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17989454 5 g 5g
17947834 10 mg 10mg
17948694 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17989454 Supplier Toronto Research Chemicals Supplier No. TRCR6848005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20283-92-5
Molecular Formula C18 H16 O8
Molecular Weight (g/mol) 360.31
Synonym Rosmarinic Acid, Benzenepropanoic acid, α-[[(2E)-3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-3,4-dihydroxy-, (αR)- (9CI), Benzenepropanoic acid, α-[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-3,4-dihydroxy-, [R-(E)]-, Cinnamic acid, 3,4-dihydroxy-, 2-ester with 3-(3,4-dihydroxyphenyl)lactic acid, Rosmarinic acid, Labiatenic acid, Labiatic acid, Mamorekku RUH 2, Meiji Red Perilla Polyphenol, NPLC 0542, RM 21A, Rosemaric acid, Rosemary acid, trans-Rosmarinic acid
IUPAC Name (2R)-3-(3,4-dihydroxyphenyl)-2-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxypropanoic acid
SMILES OC(=O)[C@@H](Cc1ccc(O)c(O)c1)OC(=O)\C=C\c2ccc(O)c(O)c2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C18H16O8/c19-12-4-1-10(7-14(12)21)3-6-17(23)26-16(18(24)25)9-11-2-5-13(20)15(22)8-11/h1-8,16,19-22H,9H2,(H,24,25)/b6-3+/t16-/m1/s1
Formula Weight 360.0845
Percent Purity >95%
Chemical Name or Material Rosmarinic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.