missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies RosetteSep™ Human Mesenchymal Stem Cell Enrichment Cocktail

Product Code. 17107681
Encompass
Click to view available options
Quantity:
200 mL bone marrow
40 mL bone marrow
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17107681

Brand: STEMCELL Technologies 15128

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunodensity isolation of untouched mesenchymal stem cells

  • The RosetteSep™ Human Mesenchymal Stem Cell Enrichment Cocktail is designed to isolate mesenchymal stem cells from fresh bone marrow by negative selection
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD3, CD14, CD19, CD38, CD66b and glycophorin A on red blood cells (RBCs)
  • When centrifuged over a buoyant density medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs
  • The purified mesenchymal stem cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium

Specifications

Cell Type Mesenchymal Stem and Progenitor Cells
For Use With (Application) Cell Isolation
Quantity 40 mL bone marrow
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.