missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RORC Monoclonal Antibody (OTI3C12), TrueMAB™, OriGene
Description
RORC Monoclonal antibody specifically detects RORC in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | RORC |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI3C12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | ZNF329 |
| Gene Accession No. | Q86UD4 |
| Gene Alias | IMD42; Nr1f3; Nuclear receptor ROR-gamma; nuclear receptor RZR-gamma; Nuclear receptor subfamily 1 group F member 3; RAR related orphan receptor C; RAR-related orphan nuclear receptor variant 2; RAR-related orphan receptor C; RAR-related orphan receptor C, isoform a; RAR-related orphan receptor gamma; retinoic acid-binding receptor gamma; retinoid-related orphan receptor gamma; retinoid-related orphan receptor-gamma; RORC; RORG; RORgamma; RZRG; RZR-GAMMA; Thor; Thymus orphan receptor; TOR |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?