Learn More
Ronidazole-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Ronidazole-d3 |
| CAS | 1015855-87-4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C6 2H3 H5 N4 O4 |
| Quantity | 50 mg |
| Synonym | Ronidazole D3, 1H-Imidazole-2-methanol','1-(methyl-d3)-5-nitro-','2-carbamate (ACI), 2-(Carbamoyloxymethyl)-1-methyl-d3-5-nitro-imidazole |
| InChI Formula | InChI=1S/C6H8N4O4/c1-9-4(3-14-6(7)11)8-2-5(9)10(12)13/h2H,3H2,1H3,(H2,7,11)/i1D3 |
| SMILES | [2H]C([2H])([2H])n1c(COC(=O)N)ncc1[N+](=O)[O-] |
| IUPAC Name | [5-nitro-1-(trideuteriomethyl)imidazol-2-yl]methyl carbamate |
| Molecular Weight (g/mol) | 203.17 |
| Show More |