missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies RoboSep™ Human Cord Blood CD34 Positive Selection Kit III
Click to view available options
Quantity:
1000 mL cord blood
Unit Size:
1 set
Description
- Isolate highly purified CD34+ cells from fresh whole umbilical cord blood (CB) using a two-step procedure
- Hematopoietic progenitor cells are pre-enriched using the RosetteSep™ Human Progenitor Cell Basic Pre-Enrichment Cocktail (15226C), with antibodies to remove cells expressing T, B and myeloid cell surface markers
- CD34+ cells are then selected using the EasySep™ Human CD34 Positive Selection Cocktail (18096C)
- f isolating CD34+ cells from whole CB in which platelet contamination is expected the EasySep™ Human Cord Blood CD34 Positive Selection Kit II (Catalog #17896) is recommended
- If isolating CD34+ cells from fresh blood or buffy coat, the Complete Kit for Human Whole Blood CD34+ Cells (Catalog #15086) is recommended
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 1000 mL cord blood |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction